accessibility menu, dialog, popup

UserName

H3COOHSOOHSOOSH

CAS RN 33007-83-9

IUPAC Name: 2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})butyl 3-sulfanylpropanoate
Synonyms: 2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})butyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 398.55
Molecular Formula: C15H26O6S3
InChi Key: IMQFZQVZKBIPCQ-UHFFFAOYSA-N
SMILES: CCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results