missing translation for 'onlineSavingsMsg'
Learn More

Trimethylhydroquinone 98.0+%, TCI America™

Catalog No. p-7136304
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T047725G 25 g
T0477100G 100 g
T0477500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T047725G Supplier TCI America Supplier No. T047725G
Only null left

Chemical Identifiers

CAS 700-13-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002346
InChI Key AUFZRCJENRSRLY-UHFFFAOYSA-N
Synonym trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
PubChem CID 12785
IUPAC Name 2,3,5-trimethylbenzene-1,4-diol
SMILES CC1=CC(=C(C(=C1O)C)C)O

Specifications

Melting Point 169°C
Quantity 25 g
UN Number 3077
Formula Weight 152.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylhydroquinone
EINECSNumber (3)-0552&(4)-0057&(9)-0913
RTECSNumber MX7820000
TSCA Yes