missing translation for 'onlineSavingsMsg'
Learn More

Trimethylethoxysilane 98.0+%, TCI America™

Supplier:  TCI America T1394250ML

 View more versions of this product

Catalog No. T1394250ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethylethoxysilane
-83°C
75°C
MFCD00009069
1993
RSIHJDGMBDPTIM-UHFFFAOYSA-N
ethoxytrimethylsilane
15772
≥98.0% (GC)
1825-62-3
Colorless
C5H14OSi
250 mL
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
CCO[Si](C)(C)C
118.25
118.25
Liquid

Chemical Identifiers

1825-62-3
118.25
RSIHJDGMBDPTIM-UHFFFAOYSA-N
15772
CCO[Si](C)(C)C
C5H14OSi
MFCD00009069
trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
ethoxytrimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2052&(2)-2053

RTECSNumber : VV4110000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions