missing translation for 'onlineSavingsMsg'
Learn More

Trimethylethoxysilane 98.0+%, TCI America™

Catalog No. p-7136754
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T139425ML 25 mL
T1394250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T139425ML Supplier TCI America Supplier No. T139425ML
Only null left

Chemical Identifiers

CAS 1825-62-3
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00009069
InChI Key RSIHJDGMBDPTIM-UHFFFAOYSA-N
Synonym trimethylethoxysilane, silane, ethoxytrimethyl, silane, trimethylethoxy, ethyl trimethylsilyl ether, trimethylsilyl ethyl ether, ethoxy trimethyl silane, trimethyl ethoxy silane, silane,ethoxytrimethyl, acmc-1btam, ethanol, tms derivative
PubChem CID 15772
IUPAC Name ethoxytrimethylsilane
SMILES CCO[Si](C)(C)C

Specifications

Melting Point -83°C
Color Colorless
Boiling Point 75°C
Quantity 25 mL
UN Number 1993
Formula Weight 118.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylethoxysilane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV4110000
TSCA Yes