missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene sulfide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 182910250

 View more versions of this product

Catalog No. AC182910250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Trimethylene sulfide
1.0280 g/mL
-2°C
1.5040 to 1.5140
25 g
1.028
XSROQCDVUIHRSI-UHFFFAOYSA-N
thietane
74.14
287-27-4
94.0°C to 94.5°C
C3H6S
MFCD00042751
17, I, 3
Solubility aq. soln.: insoluble.
C1CSC1
74.14

Chemical Identifiers

287-27-4
74.14
XSROQCDVUIHRSI-UHFFFAOYSA-N
C1CSC1
C3H6S
MFCD00042751
thietane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Highly flammable liquid and vapor.

Harmful if swallowed.

GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Store in a well-ventilated place.

Keep container tightly closed.

GHS Signal Word: Danger

EINECSNumber : 206-015-0

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only