missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene sulfide

Catalog No. p-7214338
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182910050 5 g
AC182910250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182910050 Supplier Thermo Scientific Chemicals Supplier No. 182910050
Only null left

CAS: 287-27-4 | C3H6S | 74.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 287-27-4
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00042751
InChI Key XSROQCDVUIHRSI-UHFFFAOYSA-N
IUPAC Name thietane
SMILES C1CSC1

Specifications

Density 1.0280 g/mL
Boiling Point 94.0°C to 94.5°C
Flash Point -2°C
Refractive Index 1.5040 to 1.5140
Quantity 5 g
Beilstein 17, I, 3
Specific Gravity 1.028
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 74.14
Chemical Name or Material Trimethylene sulfide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Highly flammable liquid and vapor.

Harmful if swallowed.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Store in a well-ventilated place.

Keep container tightly closed.

EINECSNumber 206-015-0

RUO – Research Use Only