Learn More
Trimethylene sulfide, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 182910050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Trimethylene sulfide | |
| 1.0280 g/mL | |
| -2°C | |
| 1.5040 to 1.5140 | |
| 5 g | |
| 1.028 | |
| XSROQCDVUIHRSI-UHFFFAOYSA-N | |
| thietane | |
| 74.14 |
| 287-27-4 | |
| 94.0°C to 94.5°C | |
| C3H6S | |
| MFCD00042751 | |
| 17, I, 3 | |
| Solubility aq. soln.: insoluble. | |
| C1CSC1 | |
| 74.14 |
Chemical Identifiers
| 287-27-4 | |
| 74.14 | |
| XSROQCDVUIHRSI-UHFFFAOYSA-N | |
| C1CSC1 |
| C3H6S | |
| MFCD00042751 | |
| thietane |
Safety and Handling
GHS H Statement Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 206-015-0
RUO â Research Use Only