missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene oxide, 97%

CAS: 503-30-0 | C3H6O | 58.08 g/mol

Supplier:  Thermo Scientific Chemicals A1445222

 View more versions of this product

Catalog No. AAA1445222


Only null left
Explore available promotions

Description

Description

Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylene oxide useful for the synthesis of 3-substituted propanols by reaction with Grignards or organolithiums in the presence of CuBr.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is hygroscopic in nature and sensitive to air. Store under inert gas.
Specifications

Specifications

Trimethylene oxide
-97°C
49°C to 50°C
C3H6O
MFCD00005167
UN1993
14,9715
Soluble in water.
C1COC1
58.08
CHEBI:30965
97%
503-30-0
0.891
−28°C (−18°F)
1.393
100 g
102382
trimethylene oxide, 1,3-propylene oxide, 1,3-epoxypropane, oxacyclobutane, cyclooxabutane, trimethylenoxid, oxetan, 1,3-trimethylene oxide, propane, 1,3-epoxy, trimethylenoxid german
AHHWIHXENZJRFG-UHFFFAOYSA-N
oxetane
10423
58.08

Chemical Identifiers

503-30-0
58.08
AHHWIHXENZJRFG-UHFFFAOYSA-N
10423
oxetane
C3H6O
MFCD00005167
trimethylene oxide, 1,3-propylene oxide, 1,3-epoxypropane, oxacyclobutane, cyclooxabutane, trimethylenoxid, oxetan, 1,3-trimethylene oxide, propane, 1,3-epoxy, trimethylenoxid german
CHEBI:30965
C1COC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H225-H302+H312+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 207-964-3

RTECSNumber : RQ6825000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only