accessibility menu, dialog, popup

UserName

CAS RN 503-30-0

IUPAC Name: oxetane
Synonyms: oxetane 1,3-trimethylene oxide trimethylenoxid 1,3-propylene oxide α,gamma-propane oxide
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: AHHWIHXENZJRFG-UHFFFAOYSA-N
SMILES: C1COC1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)