missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene Di(thiotosylate) 95.0+%, TCI America™

Catalog No. p-7127271
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23905G 5 g
D239025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23905G Supplier TCI America Supplier No. D23905G
Only null left

Chemical Identifiers

CAS 3866-79-3
Molecular Formula C17H20O4S4
Molecular Weight (g/mol) 416.583
MDL Number MFCD00014918
InChI Key QOICHLMIPNOKLN-UHFFFAOYSA-N
Synonym 1,3-Di(p-tosylthio)propane, 1,3-Propanediyl Di(thiotosylate), Trimethylene Bis(p-toluenethiosulfonate)
PubChem CID 256129
IUPAC Name 1-methyl-4-[3-(4-methylphenyl)sulfonylsulfanylpropylsulfanylsulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)SCCCSS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 416.58
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylene Di(thiotosylate) [Protecting Reagent for Active Methylene]