missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S3716

Catalog No. S-3716


Only null left
Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Trimethylamine75-50-31000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Trimethylamine
75-50-3,67-56-1
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
C3H9N
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
GETQZCLCWQTVFV-UHFFFAOYSA-N
Methanol-P and T
59.11
1146
2mL Amber Prescored Glass Ampule
0.1,99.9
0.1,99.9
MFCD00008327
Certified Reference Material
CN(C)C
N,N-dimethylmethanamine
1 mL
CHEBI:18139

Chemical Identifiers

75-50-3
59.11
GETQZCLCWQTVFV-UHFFFAOYSA-N
CHEBI:18139
CN(C)C
C3H9N
MFCD00008327
1146
N,N-dimethylmethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only