missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine N-Oxide Dihydrate 98.0+%, TCI America™

Catalog No. T0466500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0466500G 500 g
T046625G 25 g
T0466100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0466500G Supplier TCI America Supplier No. T0466500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62637-93-8
Molecular Formula C3H13NO3
Molecular Weight (g/mol) 111.14
MDL Number MFCD00149077
InChI Key PGFPZGKEDZGJQZ-UHFFFAOYSA-N
Synonym trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
PubChem CID 198430
IUPAC Name N,N-dimethylmethanamine oxide dihydrate
SMILES O.O.C[N+](C)(C)[O-]

Specifications

Melting Point 95°C
Color White
Quantity 500 g
Formula Weight 75.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine N-Oxide Dihydrate
RTECSNumber YH2850000
TSCA Yes