CAS RN 62637-93-8
IUPAC Name:
N,N-dimethylmethanamine oxide dihydrate
Synonyms:
trimethylamine oxide
Molecular Weight (g/mol):
111.14
Molecular Formula:
C3H13NO3
InChi Key:
PGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILES:
O.O.C[N+](C)(C)[O-]
1
–
1
of
1
results
1
Promotions Available
Trimethylamine N-oxide dihydrate, 98+%
Trimethylamine N-oxide dihydrate, 98+%
CAS: 62637-93-8 Molecular Formula: C3H13NO3 Molecular Weight (g/mol): 111.14 MDL Number: MFCD00149077 InChI Key: PGFPZGKEDZGJQZ-UHFFFAOYSA-N Synonym: trimethylamine-n-oxide dihydrate,trimethylamine n-oxide dihydrate,trimethylamine oxide dihydrate,trimethylamine oxide, dihydrate,n,n-dimethylmethanamine oxide, dihydrate,unii-c73wz0186w,trimethylammoniumoxid hydrat czech,methanamine oxide, n,n-dimethyl, dihydrate,methanamine, n,n-dimethyl-, n-oxide, dihydrate,n,n-dimethylmethanamine oxide dihydrate PubChem CID: 198430 IUPAC Name: N,N-dimethylmethanamine oxide;dihydrate SMILES: O.O.C[N+](C)(C)[O-]