missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine N-oxide dihydrate, 98+%

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol

Supplier:  Thermo Scientific Chemicals A1491622

 View more versions of this product

Catalog No. AAA1491622


Only null left
Explore available promotions

Description

Description

Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

Solubility
Soluble in water, ethanol, dimethyl sulfoxide and methanol. Sparingly soluble in hot chloroform. Insoluble in diethyl ether, benzene and hydrocarbon solvents.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Specifications

Specifications

Trimethylamine N-oxide dihydrate
96°C to 101°C
MFCD00149077
3612927
14,9710
Soluble in water,ethanol,dimethyl sulfoxide and methanol. Sparingly soluble in hot chloroform. Insoluble in diethyl ether,benzene and hydrocarbon solvents.
O.O.C[N+](C)(C)[O-]
111.14
111.14 (75.11 Anhydrous)
62637-93-8
C3H13NO3
100 g
Hygroscopic
trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
PGFPZGKEDZGJQZ-UHFFFAOYSA-N
N,N-dimethylmethanamine oxide;dihydrate
198430
≥98%

Chemical Identifiers

62637-93-8
111.14
PGFPZGKEDZGJQZ-UHFFFAOYSA-N
198430
O.O.C[N+](C)(C)[O-]
C3H13NO3
MFCD00149077
trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
N,N-dimethylmethanamine oxide;dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

RTECSNumber : YH2850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only