missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine N-oxide dihydrate, 98+%

Catalog No. AAA1491606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1491606 5 g
AAA1491614 25 g
AAA1491622 100 g
AAA1491636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1491606 Supplier Thermo Scientific Chemicals Supplier No. A1491606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62637-93-8 | C3H13NO3 | 111.14 g/mol

Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine N-oxide dihydrate is used as an oxidizing agent in organic synthesis. It is also used to make quaternary ammonium compounds and a warning agent for flammable gas. Further, it acts as a flavoring agent or adjuvant. It serves as an oxidant for the catalytic osmium tetraoxide cis-hydroxylation of hindered olefins. In addition to this, it is used as a reactant for C-H bond cleavage and decarbonylating agent for solvent-free reactions.

Solubility
Soluble in water, ethanol, dimethyl sulfoxide and methanol. Sparingly soluble in hot chloroform. Insoluble in diethyl ether, benzene and hydrocarbon solvents.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 62637-93-8
Molecular Formula C3H13NO3
Molecular Weight (g/mol) 111.14
MDL Number MFCD00149077
InChI Key PGFPZGKEDZGJQZ-UHFFFAOYSA-N
Synonym trimethylamine-n-oxide dihydrate, trimethylamine n-oxide dihydrate, trimethylamine oxide dihydrate, trimethylamine oxide, dihydrate, n,n-dimethylmethanamine oxide, dihydrate, unii-c73wz0186w, trimethylammoniumoxid hydrat czech, methanamine oxide, n,n-dimethyl, dihydrate, methanamine, n,n-dimethyl-, n-oxide, dihydrate, n,n-dimethylmethanamine oxide dihydrate
PubChem CID 198430
IUPAC Name N,N-dimethylmethanamine oxide;dihydrate
SMILES O.O.C[N+](C)(C)[O-]

Specifications

Melting Point 96°C to 101°C
Quantity 5 g
Beilstein 3612927
Sensitivity Hygroscopic
Merck Index 14,9710
Solubility Information Soluble in water,ethanol,dimethyl sulfoxide and methanol. Sparingly soluble in hot chloroform. Insoluble in diethyl ether,benzene and hydrocarbon solvents.
Formula Weight 111.14 (75.11 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Trimethylamine N-oxide dihydrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber YH2850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only