missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic anhydride, 99%

CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol

Supplier:  Thermo Scientific Chemicals B2298322

 View more versions of this product

Catalog No. AAB2298322


Only null left
Explore available promotions

Description

Description

Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

Solubility
Miscible with acetonitrile.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications

Specifications

Trimethylacetic anhydride
0.918
63°C (145°F)
C10H18O3
((CH3)3CCO)2O
100 g
386552
pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
PGZVFRAEAAXREB-UHFFFAOYSA-N
2,2-dimethylpropanoyl 2,2-dimethylpropanoate
15234
99%
1538-75-6
193°C
Unpleasant
1.409
MFCD00008842
UN3265
Moisture sensitive
Miscible with acetonitrile.
CC(C)(C)C(=O)OC(=O)C(C)(C)C
186.25
186.25

Chemical Identifiers

1538-75-6
186.25
PGZVFRAEAAXREB-UHFFFAOYSA-N
15234
CC(C)(C)C(=O)OC(=O)C(C)(C)C
C10H18O3
MFCD00008842
pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
2,2-dimethylpropanoyl 2,2-dimethylpropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 216-263-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only