missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic anhydride, 99%

Catalog No. p-7026773
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2298314 25 g
AAB2298322 100 g
AAB2298336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2298314 Supplier Thermo Scientific Chemicals Supplier No. B2298314
Only null left

CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol

Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetic anhydride is used as acylation and esterification reagents. It is involved in acylation and esterification reactions with anilines and phenols. It is also used in solid-phase oligonucleotide synthesis and in the kinetic resolution of racemic 2-hydroxy-gamma-butyrolactones with diphenylacetic acid. It is also employed in the production of cyano-4, N-tert-butyloxycarbonyl piperidine.

Solubility
Miscible with acetonitrile.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 1538-75-6
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00008842
InChI Key PGZVFRAEAAXREB-UHFFFAOYSA-N
Synonym pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
PubChem CID 15234
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C

Specifications

Density 0.918
Boiling Point 193°C
Flash Point 63°C (145°F)
Odor Unpleasant
Refractive Index 1.409
Linear Formula ((CH3)3CCO)2O
Quantity 25 g
UN Number UN3265
Beilstein 386552
Sensitivity Moisture sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 186.25
Percent Purity 99%
Chemical Name or Material Trimethylacetic anhydride
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 216-263-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only