missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetaldehyde, 95%

CAS: 630-19-3 | C5H10O | 86.13 g/mol

Supplier:  Thermo Scientific Chemicals A1501314

 View more versions of this product

Catalog No. AAA1501314

Only null left
Explore available promotions

Description

Description

Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is virtually insoluble in water but soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from oxidizing agents, strong reducing agents.
Specifications

Specifications

Trimethylacetaldehyde
6°C
74°C to 76°C
Pleasant
1.3794
MFCD00006962
UN1989
Air sensitive
It is virtually insoluble in water but soluble in chloroform.
CC(C)(C)C=O
86.13
86.13
630-19-3
0.775
−15°C (5°F)
C5H10O
(CH3)3CCHO
25 g
506060
trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
FJJYHTVHBVXEEQ-UHFFFAOYSA-N
2,2-dimethylpropanal
12417
96%

Chemical Identifiers

630-19-3
86.13
FJJYHTVHBVXEEQ-UHFFFAOYSA-N
12417
CC(C)(C)C=O
C5H10O
MFCD00006962
trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
2,2-dimethylpropanal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 211-134-6

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only