accessibility menu, dialog, popup

UserName

CH3CH3H3CO

CAS RN 630-19-3

IUPAC Name: 2,2-dimethylpropanal
Synonyms: pivaldehyde pivalaldehyde
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: FJJYHTVHBVXEEQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C=O
Boiling Point 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (2)
  • (4)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)