missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl(vinyloxy)silane 95.0+%, TCI America™
$59.65 - $182.68
Chemical Identifiers
| CAS | 6213-94-1 |
|---|---|
| Molecular Formula | C5H12OSi |
| Molecular Weight (g/mol) | 116.235 |
| MDL Number | MFCD00054764 |
| InChI Key | HFTNNOZFRQLFQB-UHFFFAOYSA-N |
| Synonym | trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether |
| PubChem CID | 80344 |
| IUPAC Name | ethenoxy(trimethyl)silane |
| SMILES | C[Si](C)(C)OC=C |
Chemical Identifiers
| 6213-94-1 | |
| 116.235 | |
| HFTNNOZFRQLFQB-UHFFFAOYSA-N | |
| 80344 | |
| C[Si](C)(C)OC=C |
| C5H12OSi | |
| MFCD00054764 | |
| trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether | |
| ethenoxy(trimethyl)silane |
Specifications
| 6213-94-1 | |
| 75°C | |
| MFCD00054764 | |
| 1993 | |
| HFTNNOZFRQLFQB-UHFFFAOYSA-N | |
| ethenoxy(trimethyl)silane | |
| 80344 | |
| ≥95.0% (GC) | |
| Trimethyl(vinyloxy)silane |
| Yellow | |
| C5H12OSi | |
| 1 g | |
| trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether | |
| C[Si](C)(C)OC=C | |
| 116.235 | |
| 116.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2960
TSCA : Yes
Recommended Storage : Refrigerator