missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(vinyloxy)silane 95.0+%, TCI America™

Catalog No. T25611G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25611G 1 g
T25615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T25611G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T25611G

Chemical Identifiers

CAS 6213-94-1
Molecular Formula C5H12OSi
Molecular Weight (g/mol) 116.235
MDL Number MFCD00054764
InChI Key HFTNNOZFRQLFQB-UHFFFAOYSA-N
Synonym trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether
PubChem CID 80344
IUPAC Name ethenoxy(trimethyl)silane
SMILES C[Si](C)(C)OC=C

Specifications

Color Yellow
Boiling Point 75°C
Quantity 1 g
UN Number 1993
Formula Weight 116.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(vinyloxy)silane
EINECSNumber (2)-2960
TSCA Yes
Recommended Storage Refrigerator