missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(tridecafluorohexyl)silane 97.0+%, TCI America™

Catalog No. T35951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35951G 1 g
T35955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35951G Supplier TCI America Supplier No. T35951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135841-49-5
Molecular Formula C9H9F13Si
Molecular Weight (g/mol) 392.24
MDL Number MFCD20489406
InChI Key AHGSEBBCGYXEBF-UHFFFAOYSA-N
Synonym Trimethyl(perfluorohexyl)silane
PubChem CID 10894469
IUPAC Name trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)silane
SMILES C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 392.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(tridecafluorohexyl)silane