missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl trans-Aconitate 97.0+%, TCI America™

Catalog No. A07255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07255G 5 g
A072525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07255G Supplier TCI America Supplier No. A07255G

Chemical Identifiers

CAS 4271-99-2
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00059304
InChI Key DZAIBGWGBBQGPZ-GQCTYLIASA-N
Synonym trans-Aconitic Acid Trimethyl Ester
PubChem CID 6376716
IUPAC Name trimethyl (E)-prop-1-ene-1,2,3-tricarboxylate
SMILES COC(=O)CC(=CC(=O)OC)C(=O)OC

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl trans-Aconitate