accessibility menu, dialog, popup

UserName

CAS RN 4271-99-2

CH3OOOOCH3OOCH3(E/Z)

CAS RN 4271-99-2

IUPAC Name: 1,2,3-trimethyl prop-1-ene-1,2,3-tricarboxylate
Synonyms: 1,2,3-trimethyl prop-1-ene-1,2,3-tricarboxylate
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: DZAIBGWGBBQGPZ-UHFFFAOYSA-N
SMILES: COC(=O)CC(=CC(=O)OC)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results