missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl phosphite, 97%, Thermo Scientific Chemicals

Catalog No. p-7049830
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3013222 100 g
AA3013236 500 g
AA30132A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3013222 Supplier Thermo Scientific Chemicals Supplier No. 03013222
Only null left

It is an important raw material and intermediate used in Organic synthesis..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-45-9
Molecular Formula C3H9O3P
Molecular Weight (g/mol) 124.076
MDL Number MFCD00008350
InChI Key CYTQBVOFDCPGCX-UHFFFAOYSA-N
Synonym trimethoxyphosphine, phosphorous acid, trimethyl ester, trimethylphosphite, methyl phosphite, trimethylfosfit, trimethoxyfosfin, trimethylfosfit czech, trimethoxyfosfin czech, fosforyn trojmetylowy czech, unii-26q0321zdg
PubChem CID 8472
IUPAC Name trimethyl phosphite
SMILES COP(OC)OC

Specifications

Melting Point °C to 78°C
Density 1.052 g/mL
Boiling Point 111°C to 112°C
Flash Point 27°C (80°F)
Assay Percent Range 97%
Refractive Index 1.408
Linear Formula (CH3O)3P
Quantity 100 g
UN Number UN2329
Sensitivity Air and moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 124.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethyl phosphite
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H226-H361-H302-H315-H319-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIMETHYL PHOSPHITE
EINECSNumber 204-471-5
RTECSNumber TH1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only