missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl(pentafluorophenyl)silane 98.0+%, TCI America™
Supplier: TCI America T30125G
Specifications
| Trimethyl(pentafluorophenyl)silane | |
| Colorless | |
| C9H9F5Si | |
| 5 g | |
| trimethyl pentafluorophenyl silane, trimethyl perfluorophenyl silane, pentafluorophenyltrimethylsilane, pentafluorophenyl trimethyl silane, silane, trimethyl pentafluorophenyl, trimethylsilylpentafluorobenzene, pentafluorophenyl-trimethylsilane, pentafluorophenyl trimethylsilane, trimethyl-2,3,4,5,6-pentafluorophenyl silane | |
| C[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 240.25 | |
| 240.25 | |
| Liquid |
| 1206-46-8 | |
| 165°C | |
| MFCD00092630 | |
| 1993 | |
| GABHTFORECKGBB-UHFFFAOYSA-N | |
| trimethyl(2,3,4,5,6-pentafluorophenyl)silane | |
| 297554 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1206-46-8 | |
| 240.25 | |
| GABHTFORECKGBB-UHFFFAOYSA-N | |
| 297554 | |
| C[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F |
| C9H9F5Si | |
| MFCD00092630 | |
| trimethyl pentafluorophenyl silane, trimethyl perfluorophenyl silane, pentafluorophenyltrimethylsilane, pentafluorophenyl trimethyl silane, silane, trimethyl pentafluorophenyl, trimethylsilylpentafluorobenzene, pentafluorophenyl-trimethylsilane, pentafluorophenyl trimethylsilane, trimethyl-2,3,4,5,6-pentafluorophenyl silane | |
| trimethyl(2,3,4,5,6-pentafluorophenyl)silane |
Safety and Handling
TSCA : No