missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(pentafluorophenyl)silane 98.0+%, TCI America™

Supplier:  TCI America T30121G

 View more versions of this product

Catalog No. T30121G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethyl(pentafluorophenyl)silane
Colorless
C9H9F5Si
1 g
trimethyl pentafluorophenyl silane, trimethyl perfluorophenyl silane, pentafluorophenyltrimethylsilane, pentafluorophenyl trimethyl silane, silane, trimethyl pentafluorophenyl, trimethylsilylpentafluorobenzene, pentafluorophenyl-trimethylsilane, pentafluorophenyl trimethylsilane, trimethyl-2,3,4,5,6-pentafluorophenyl silane
C[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F
240.25
240.25
Liquid
1206-46-8
165°C
MFCD00092630
1993
GABHTFORECKGBB-UHFFFAOYSA-N
trimethyl(2,3,4,5,6-pentafluorophenyl)silane
297554
≥98.0% (GC)

Chemical Identifiers

1206-46-8
240.25
GABHTFORECKGBB-UHFFFAOYSA-N
297554
C[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F
C9H9F5Si
MFCD00092630
trimethyl pentafluorophenyl silane, trimethyl perfluorophenyl silane, pentafluorophenyltrimethylsilane, pentafluorophenyl trimethyl silane, silane, trimethyl pentafluorophenyl, trimethylsilylpentafluorobenzene, pentafluorophenyl-trimethylsilane, pentafluorophenyl trimethylsilane, trimethyl-2,3,4,5,6-pentafluorophenyl silane
trimethyl(2,3,4,5,6-pentafluorophenyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions