missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthobenzoate 95.0+%, TCI America™

Catalog No. T22875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22875G 5 g
T228725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22875G Supplier TCI America Supplier No. T22875G

Chemical Identifiers

CAS 707-07-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008474
InChI Key IECKAVQTURBPON-UHFFFAOYSA-N
Synonym trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate
PubChem CID 69720
IUPAC Name trimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)(OC)OC

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthobenzoate