Learn More
Trimethyl orthobenzoate, 98%
CAS: 707-07-3 | C10H14O3 | 182.219 g/mol
$81.56 - $956.79
Chemical Identifiers
| CAS | 707-07-3 |
|---|---|
| Molecular Formula | C10H14O3 |
| Molecular Weight (g/mol) | 182.219 |
| MDL Number | MFCD00008474 |
| InChI Key | IECKAVQTURBPON-UHFFFAOYSA-N |
| Synonym | trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate |
| PubChem CID | 69720 |
| IUPAC Name | trimethoxymethylbenzene |
| SMILES | COC(C1=CC=CC=C1)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1027009
|
Thermo Scientific Chemicals
A1027009 |
10 g |
Each for $81.56
|
|
|||||
|
AAA1027018
|
Thermo Scientific Chemicals
A1027018 |
50 g |
Each for $238.32
|
|
|||||
|
AAA1027030
|
Thermo Scientific Chemicals
A1027030 |
250 g |
Each for $956.79
|
|
|||||
Description
Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.
Solubility
Soluble in organic solvents.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with oxidizing agents and moisture.
Chemical Identifiers
| 707-07-3 | |
| 182.219 | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| 69720 | |
| COC(C1=CC=CC=C1)(OC)OC |
| C10H14O3 | |
| MFCD00008474 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| trimethoxymethylbenzene |
Specifications
| 707-07-3 | |
| 87°C to 88°C (7 mmHg) | |
| C10H14O3 | |
| MFCD00008474 | |
| 777623 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| trimethoxymethylbenzene | |
| 69720 | |
| 98% |
| 1.06 | |
| 82°C (179°F) | |
| 1.489 | |
| 10 g | |
| Moisture sensitive | |
| Soluble in organic solvents. | |
| COC(C1=CC=CC=C1)(OC)OC | |
| 182.219 | |
| 182.22 | |
| Trimethyl orthobenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 211-897-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only