missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(nonafluorobutyl)silane 98.0+%, TCI America™

Catalog No. T35941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35941G 1 g
T35945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35941G Supplier TCI America Supplier No. T35941G
Only null left

Chemical Identifiers

CAS 204316-01-8
Molecular Formula C7H9F9Si
Molecular Weight (g/mol) 292.22
MDL Number MFCD20489405
InChI Key YHSOLWOJHMOEFX-UHFFFAOYSA-N
Synonym Trimethyl(perfluorobutyl)silane
PubChem CID 15534208
IUPAC Name trimethyl(1,1,2,2,3,3,4,4,4-nonafluorobutyl)silane
SMILES C[Si](C)(C)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 100°C
Quantity 1 g
UN Number 1993
Formula Weight 292.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(nonafluorobutyl)silane