CAS RN 204316-01-8
IUPAC Name:
trimethyl(1,1,2,2,3,3,4,4,4-nonafluorobutyl)silane
Synonyms:
trimethyl(1,1,2,2,3,3,4,4,4-nonafluorobutyl)silane
Molecular Weight (g/mol):
292.22
Molecular Formula:
C7H9F9Si
InChi Key:
YHSOLWOJHMOEFX-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Trimethyl(nonafluorobutyl)silane 98.0+%, TCI America™
CAS: 204316-01-8 Molecular Formula: C7H9F9Si Molecular Weight (g/mol): 292.22 MDL Number: MFCD20489405 InChI Key: YHSOLWOJHMOEFX-UHFFFAOYSA-N Synonym: Trimethyl(perfluorobutyl)silane PubChem CID: 15534208 IUPAC Name: trimethyl(1,1,2,2,3,3,4,4,4-nonafluorobutyl)silane SMILES: C[Si](C)(C)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F