missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl citrate, Thermo Scientific™

Chemical Identifiers

CAS 1587-20-8
Molecular Formula C9H14O7
Molecular Weight (g/mol) 234.2
InChI Key HDDLVZWGOPWKFW-UHFFFAOYSA-N
PubChem CID 74112
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC462250050
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC462250250
View Documents Promotion Details
N/A 25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1587-20-8
234.2
74112
C9H14O7
HDDLVZWGOPWKFW-UHFFFAOYSA-N

Specifications

1587-20-8
C9H14O7
HDDLVZWGOPWKFW-UHFFFAOYSA-N
74112
Trimethyl citrate
176°C (16.0mmHg)
Solubility in water: .. Other solubilities: soluble in chloroform,methanol
234.2
234.2
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-449-2

SDS
Documents

Documents

RUO – Research Use Only