missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl citrate, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 462250050

Quantity 5g

 View more versions of this product

Catalog No. AC462250050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1587-20-8
234.2
74112
C9H14O7
HDDLVZWGOPWKFW-UHFFFAOYSA-N

Specifications

Trimethyl citrate
176°C (16.0mmHg)
5g
HDDLVZWGOPWKFW-UHFFFAOYSA-N
74112
1587-20-8
C9H14O7
Solubility in water: .. Other solubilities: soluble in chloroform,methanol
234.2
234.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-449-2

SDS
Documents

Documents

Promotions

Promotions