missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl Borate 98.0+%, TCI America™
Supplier: TCI America B0522100ML
Specifications
| Trimethyl Borate | |
| -29°C | |
| 68°C | |
| MFCD00008346 | |
| 2416 | |
| WRECIMRULFAWHA-UHFFFAOYSA-N | |
| trimethyl borate | |
| 8470 | |
| 103.91 | |
| Liquid |
| 121-43-7 | |
| Yellow | |
| C3H9BO3 | |
| 100 mL | |
| trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3 | |
| B(OC)(OC)OC | |
| 103.912 | |
| CHEBI:38913 | |
| ≥98.0% (T) |
Chemical Identifiers
| 121-43-7 | |
| 103.912 | |
| WRECIMRULFAWHA-UHFFFAOYSA-N | |
| 8470 | |
| trimethyl borate |
| C3H9BO3 | |
| MFCD00008346 | |
| trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3 | |
| CHEBI:38913 | |
| B(OC)(OC)OC |
Safety and Handling
EINECSNumber : (2)-2101&(2)-2114
RTECSNumber : ED5600000
TSCA : Yes