missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl[3-(triethoxysilyl)propyl]ammonium Chloride 98.0+%, TCI America™

Catalog No. T22465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22465G 5 g
T224625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22465G Supplier TCI America Supplier No. T22465G
Only null left

Chemical Identifiers

CAS 84901-27-9
Molecular Formula C12H30ClNO3Si
Molecular Weight (g/mol) 299.91
MDL Number MFCD00043316
InChI Key MVYLHRSUMWTUJE-UHFFFAOYNA-M
PubChem CID 23002981
IUPAC Name trimethyl[1-(triethoxysilyl)propyl]azanium chloride
SMILES [Cl-].CCO[Si](OCC)(OCC)C(CC)[N+](C)(C)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 299.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl[3-(triethoxysilyl)propyl]ammonium Chloride
EINECSNumber (2)-2094&(2)-2095
TSCA No