accessibility menu, dialog, popup

UserName

ClH3COSiOCH3OH3CCH3NCH3CH3H3C

CAS RN 84901-27-9

IUPAC Name: trimethyl[1-(triethoxysilyl)propyl]azanium chloride
Synonyms: trimethyl[1-(triethoxysilyl)propyl]azanium chloride
Molecular Weight (g/mol): 299.91
Molecular Formula: C12H30ClNO3Si
InChi Key: MVYLHRSUMWTUJE-UHFFFAOYNA-M
SMILES: [Cl-].CCO[Si](OCC)(OCC)C(CC)[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results