missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxyboroxine

Catalog No. p-7080499
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8915314 25 g
AA8915330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8915314 Supplier Thermo Scientific Chemicals Supplier No. 08915314

CAS: 102-24-9 | C3H9B3O6 | 173.53 g/mol

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-24-9
Molecular Formula C3H9B3O6
Molecular Weight (g/mol) 173.53
MDL Number MFCD00005946
InChI Key ZFMOJHVRFMOIGF-UHFFFAOYSA-N
Synonym trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
PubChem CID 66880
SMILES COB1OB(OC)OB(OC)O1

Specifications

Melting Point 10°C to 11°C
Density 1.216 g/mL
Boiling Point 130°C
Flash Point 14°C (57°F)
Odor Strong
Refractive Index 1.3996
Quantity 25 g
UN Number UN2924
Beilstein 1709158
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name trimethoxy-1,3,5,2,4,6-trioxatriborinane
Formula Weight 173.53
Physical Form Liquid
Chemical Name or Material Trimethoxyboroxine
Show More Show Less
Hazard Category H225-H315-H318
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P310-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 203-016-8
RTECSNumber ED8720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only