missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy(propyl)silane 98.0+%, TCI America™
$57.75 - $192.52
Chemical Identifiers
| CAS | 1067-25-0 |
|---|---|
| Molecular Formula | C6H16O3Si |
| Molecular Weight (g/mol) | 164.28 |
| MDL Number | MFCD00043026 |
| InChI Key | HQYALQRYBUJWDH-UHFFFAOYSA-N |
| Synonym | trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane |
| PubChem CID | 61254 |
| IUPAC Name | trimethoxy(propyl)silane |
| SMILES | CCC[Si](OC)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T180125ML
|
TCI America
T180125ML |
25 mL |
Each for $57.75
|
|
|||||
|
T1801500ML
|
TCI America
T1801500ML |
500 mL |
Each for $192.52
|
|
|||||
Chemical Identifiers
| 1067-25-0 | |
| 164.28 | |
| HQYALQRYBUJWDH-UHFFFAOYSA-N | |
| 61254 | |
| CCC[Si](OC)(OC)OC |
| C6H16O3Si | |
| MFCD00043026 | |
| trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane | |
| trimethoxy(propyl)silane |
Specifications
| 1067-25-0 | |
| C6H16O3Si | |
| 25 mL | |
| trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane | |
| CCC[Si](OC)(OC)OC | |
| 164.28 | |
| 164.28 | |
| Liquid |
| Colorless | |
| MFCD00043026 | |
| 1993 | |
| HQYALQRYBUJWDH-UHFFFAOYSA-N | |
| trimethoxy(propyl)silane | |
| 61254 | |
| ≥98.0% (GC) | |
| Trimethoxy(propyl)silane |
Safety and Handling
EINECSNumber : (2)-2052&(2)-2053
TSCA : Yes