missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxy(propyl)silane 98.0+%, TCI America™

Catalog No. t1801500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
T1801500ML 500 mL
T180125ML 25 mL
2 options

Catalog No. T1801500ML

Supplier: TCI America T1801500ML

Only null left

Chemical Identifiers

CAS 1067-25-0
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00043026
InChI Key HQYALQRYBUJWDH-UHFFFAOYSA-N
Synonym trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane
PubChem CID 61254
IUPAC Name trimethoxy(propyl)silane
SMILES CCC[Si](OC)(OC)OC

Specifications

Color Colorless
Quantity 500 mL
UN Number 1993
Formula Weight 164.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(propyl)silane
EINECSNumber (2)-2052&(2)-2053
TSCA Yes