missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy(pentafluorophenyl)silane 97.0+%, TCI America™
$242.33 - $726.98
Chemical Identifiers
| CAS | 223668-64-2 |
|---|---|
| Molecular Formula | C9H9F5O3Si |
| Molecular Weight (g/mol) | 288.245 |
| MDL Number | MFCD04039205 |
| InChI Key | XFFHTZIRHGKTBQ-UHFFFAOYSA-N |
| PubChem CID | 2782640 |
| IUPAC Name | trimethoxy-(2,3,4,5,6-pentafluorophenyl)silane |
| SMILES | CO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
Chemical Identifiers
| 223668-64-2 | |
| 288.245 | |
| XFFHTZIRHGKTBQ-UHFFFAOYSA-N | |
| trimethoxy-(2,3,4,5,6-pentafluorophenyl)silane |
| C9H9F5O3Si | |
| MFCD04039205 | |
| 2782640 | |
| CO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC |
Specifications
| 223668-64-2 | |
| C9H9F5O3Si | |
| 1 g | |
| CO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC | |
| 288.245 | |
| 288.25 | |
| Liquid |
| Colorless | |
| MFCD04039205 | |
| XFFHTZIRHGKTBQ-UHFFFAOYSA-N | |
| trimethoxy-(2,3,4,5,6-pentafluorophenyl)silane | |
| 2782640 | |
| ≥97.0% (GC) | |
| Trimethoxy(pentafluorophenyl)silane |
Safety and Handling
TSCA : No