accessibility menu, dialog, popup

UserName

CAS RN 223668-64-2

H3COSiFFFFFOH3COCH3

CAS RN 223668-64-2

IUPAC Name: trimethoxy(2,3,4,5,6-pentafluorophenyl)silane
Synonyms: trimethoxy(2,3,4,5,6-pentafluorophenyl)silane
Molecular Weight (g/mol): 288.25
Molecular Formula: C9H9F5O3Si
InChi Key: XFFHTZIRHGKTBQ-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results