missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trimethoprim, 98%

Catalog No. p-7077531
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455120050 5 g
AC455120250 25 g
AC455121000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455120050 Supplier Thermo Scientific Chemicals Supplier No. 455120050

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial. | CAS: 738-70-5 | C14H18N4O3 | 290.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced • Trimethoprim has antibacterial and antiprotozoal properties

Applications

• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects

• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.32
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym trimethoprim, proloprim, trimpex, bactramin, monotrim, monotrimin, trimetoprim, trimopan, monoprim, syraprim
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Melting Point 234.0°C to 236.0°C
Loss on Drying 0.5% max. (105°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 290.32
Percent Purity 98%
Chemical Name or Material Trimethoprim
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 212-006-2

RUO – Research Use Only