missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Trimethoprim

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial. | CAS: 738-70-5 | C14H18N4O3

Supplier:  Thermo Scientific Chemicals J6305314

 View more versions of this product

Catalog No. AAJ6305314


Only null left

Description

Description

An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.Trimethoprim is an antibiotic involved in the treatment of urinary tract infections, middle ear infections and traveler’s diarrhea. It is associated with sulfamethoxazole and interferes with the cellular metabolism of folic acid in the bacterial cell by blocking the biosynthesis of nucleotides. Furthermore, It is also used to treat and prevent Pneumocystis jiroveci pneumonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced
  • Trimethoprim has antibacterial and antiprotozoal properties

Applications

  • Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects
  • Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression
Specifications

Specifications

25 g
2°C to 8°C
738-70-5
290.32g/mol
199°C to 203°C
Signal Transduction Reagent
625127
COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
290.32
White
An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
C14H18N4O3
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
Powder
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine

Chemical Identifiers

738-70-5
C14H18N4O3
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-006-2

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only