missing translation for 'onlineSavingsMsg'
Learn More

Trimeprazine Tartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018652
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018652 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018652 Supplier Merck Emd Millipore Supplier No. PHR17491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trimeprazine Tartrate belongs to the group of phenothiazine derivatives, which generally exhibit antipsychotropic, anticholinergic and antihistaminic activities. It is widely used for the treatment of moderate and severe mental and emotional conditions.

Specifications

CAS 4330-99-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H22N2 S · 0.5C4H6O6
Synonym Alimemazine hemitartrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 373.49
Grade Certified Reference Material