missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic Anhydride Chloride 98.0+%, TCI America™

Supplier:  TCI America T1079100G

 View more versions of this product

Catalog No. T1079100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimellitic Anhydride Chloride
69°C
195°C
MFCD00005924
3261
NJMOHBDCGXJLNJ-UHFFFAOYSA-N
1,3-dioxo-2-benzofuran-5-carbonyl chloride
70998
≥98.0% (T)
1204-28-0
White
C9H3ClO4
100 g
trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride
C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O
210.569
210.57
Crystalline Powder

Chemical Identifiers

1204-28-0
210.569
NJMOHBDCGXJLNJ-UHFFFAOYSA-N
70998
C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O
C9H3ClO4
MFCD00005924
trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride
1,3-dioxo-2-benzofuran-5-carbonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1361

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions