accessibility menu, dialog, popup

UserName

ClOOOO

CAS RN 1204-28-0

IUPAC Name: 1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl chloride
Synonyms: trimellitic anhydride chloride
Molecular Weight (g/mol): 210.57
Molecular Formula: C9H3ClO4
InChi Key: NJMOHBDCGXJLNJ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)