missing translation for 'onlineSavingsMsg'
Learn More

Trimellitic Anhydride Chloride 98.0+%, TCI America™

Catalog No. T1079500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1079500G 500 g
T107925G 25 g
T1079100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1079500G Supplier TCI America Supplier No. T1079500G

Chemical Identifiers

CAS 1204-28-0
Molecular Formula C9H3ClO4
Molecular Weight (g/mol) 210.569
MDL Number MFCD00005924
InChI Key NJMOHBDCGXJLNJ-UHFFFAOYSA-N
Synonym trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride
PubChem CID 70998
IUPAC Name 1,3-dioxo-2-benzofuran-5-carbonyl chloride
SMILES C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O

Specifications

Melting Point 69°C
Color White
Boiling Point 195°C
Quantity 500 g
UN Number 3261
Formula Weight 210.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimellitic Anhydride Chloride
EINECSNumber (3)-1361
TSCA Yes