missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM5 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520047
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520047 Supplier Invitrogen™ Supplier No. PA520047
Only null left

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~35kDa. A suggested positive control is human bladder tissue lysate. PA5-20047 can be used with blocking peptide PEP-0166.

TRIM5 is a member of a broad family of otherwise unrelated proteins defined by the presence of a tripartite motif containing a RING domain, a B-box type 1, and a B-box type 2, followed by a coiled-coil region. TRIM5 has five alternately spliced isoforms, the longest of which is the alpha variant which also contains a carboxy-terminal B30.2 (SPRY) domain. While a function has not yet been assigned to TRIM5-gamma, it is known that expression of TRIM5-a variants from humans, rhesus monkeys, and African green monkeys enable resistance to infection by various retroviruses including HIV-1, albeit at differing efficiencies. Furthermore, the TRIM5δ isoform appears to serve as a scaffold for the assembly of endogenous BTBD1 and BTBD2 proteins and also exhibits autoubiquitination activity in a RING finger- and UbcH5B-dependent manner.

Specifications

Antigen TRIM5 gamma
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRIM5
Gene Accession No. Q9C035
Gene Alias RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing protein 5
Gene Symbols TRIM5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids near the carboxy terminus of human TRIM5 gamma.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85363
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less