missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM34 Monoclonal Antibody (K24023_3C6), Invitrogen™

Catalog No. MA556308
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556308 200 μL
MA555928 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556308 Supplier Invitrogen™ Supplier No. MA556308
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GEKLLLFCKE DRKVICWLCE RSQEHRGHHT VLTEEVFKEC QEKLQAVLKR LKKEEEEAEK LEADIREEKT SWKYQVQTER QRIQTEFDQL RSILNNEEQR ELQRLEEEEK KTLDKFAEAE

TRIM34 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Expression of this gene is up-regulated by interferon. This gene is mappped to chromosome 11p15, where it resides within a TRIM gene cluster. Alternate splicing of this gene generates four transcript variants. Additionally, a read-through transcript transcribed from this gene and TRIM6 has been observed.

Specifications

Antigen TRIM34
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K24023_3C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TRIM34
Gene Accession No. Q9BYJ4
Gene Alias IFP1; Interferon-responsive finger protein 1; RING finger protein 21; ring finger protein 21, interferon-responsive; RNF21; TRIM34; tripartite motif containing 34; tripartite motif-containing 34; tripartite motif-containing protein 34
Gene Symbols TRIM34
Host Species Mouse
Immunogen Recombinant human TRIM34 (immunogen range is 101-220).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less