missing translation for 'onlineSavingsMsg'
Learn More

TRIM33/TIF1gamma Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559136
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501559136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501559136 Supplier Bethyl Laboratories Inc Supplier No. A301058A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

The protein encoded by TIF1 gamma is thought to be a transcriptional corepressor. However, molecules that interact with this protein have not yet been identified. The protein is a member of the tripartite motif family. This motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Three alternatively spliced transcript variants for TIF1 gamma have been described, however, the full-length nature of one variant has not been determined.

Specifications

Antigen TRIM33/TIF1gamma
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TRIM33
Gene Accession No. Q9UPN9
Gene Alias ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA, TRIM33, tripartite motif containing 33
Gene Symbols TRIM33
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less